EMAIL THIS PAGE TO A FRIEND
Psychopharmacology Series
The effect of chronic phenytoin administration on single prolonged stress induced extinction retention deficits and glucocorticoid upregulation in the rat medial prefrontal cortex.
PMID 24879497