EMAIL THIS PAGE TO A FRIEND
Environmental Toxicology and Pharmacology
Viola mandshurica ethanolic extract prevents high-fat-diet-induced obesity in mice by activating AMP-activated protein kinase.
PMID 24879516