EMAIL THIS PAGE TO A FRIEND
Cancer letters
XMD8-92 inhibits pancreatic tumor xenograft growth via a DCLK1-dependent mechanism.
PMID 24880079
Abstract
XMD8-92 is a kinase inhibitor with anti-cancer activity against lung and cervical cancers, but its effect on pancreatic ductal adenocarcinoma (PDAC) remains unknown. Doublecortin-like kinase1 (DCLK1) is upregulated in various cancers including PDAC. In this study, we showed that XMD8-92 inhibits AsPC-1 cancer cell proliferation and tumor xenograft growth. XMD8-92 treated tumors demonstrated significant downregulation of DCLK1 and several of its downstream targets (including c-MYC, KRAS, NOTCH1, ZEB1, ZEB2, SNAIL, SLUG, OCT4, SOX2, NANOG, KLF4, LIN28, VEGFR1, and VEGFR2) via upregulation of tumor suppressor miRNAs let-7a, miR-144, miR-200a-c, and miR-143/145; it did not however affect BMK1 downstream genes p21 and p53. These data taken together suggest that XMD8-92 treatment results in inhibition of DCLK1 and downstream oncogenic pathways (EMT, pluripotency, angiogenesis and anti-apoptotic), and is a promising chemotherapeutic agent against PDAC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1382
XMD8-92, ≥98% (HPLC)
C26H30N6O3