EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Transient GFER knockdown in vivo impairs liver regeneration after partial hepatectomy.
PMID 24880092
Abstract
Augmenter of Liver Regeneration is a protein encoded by the Growth Factor Erv1-Like gene. Its biological properties are crucial for cell survival since knock-out mice for Growth Factor Erv1-Like gene do not survive. In this study, we injected hepatotropic adenoviral particles harboring oligonucleotide sequences against Growth Factor Erv1-Like gene into 70% partially hepatectomized rats and studied the effect of gene silencing on the progression liver regeneration. Partially hepatectomized rats were divided into three groups of animals and, before surgery, received either phosphate buffer saline, or adenoviral particles alone or adenoviral particles harboring the oligonucleotide silencing sequence. In each group, rats were sacrificed at 12, 24 and 48 h after surgery. Liver tissues were collected to analyze the expression of Augmenter of Liver Regeneration, Bax, Bcl-2 and activated Caspase-9 and -3, as well as hepatocyte proliferation and apoptosis, polyamines levels and histological and ultrastructural features. Growth Factor Erv1-Like gene silencing reduced the compensatory hepatocellular proliferation triggered by surgery through (i) the reduction of polyamines synthesis, hepatocyte proliferation and anti-apoptotic gene expression and (ii) the increase of pro-apoptotic gene expression and caspase activation. For the first time, using a technique of gene silencing in vivo, our results demonstrate that Growth Factor Erv1-Like gene knock-down, i.e., the lack of Augmenter of Liver Regeneration, modifies the expression of genes involved in cell apoptosis and inhibits early phase of DNA synthesis. As a consequence, a promotion of cell death and a reduction of cell proliferation occurs.