EMAIL THIS PAGE TO A FRIEND
Chemosphere
Persistent halogenated compounds in captive Chinese alligators (Alligator sinensis) from China.
PMID 24880595
Abstract
While a number of studies have reported residual levels of persistent halogenated compounds (PHCs) in crocodilia, there is still a dearth of information on the Chinese alligator, a critically endangered crocodilian species. In the present study, several PHCs, including polybrominated diphenyl ethers (PBDEs), polychlorinated biphenyls (PCBs), and dichlorodiphenyltrichloroethane and its metabolites (DDTs), were detected in the adult tissues, neonates, and eggs of captive Chinese alligators from China. The concentrations of ΣPBDEs, ΣPCBs, and ΣDDTs in Chinese alligators ranged from 0.11 to 16.1, 1.12 to 22.2, and 6.03 to 1020ngg(-1) wet weight, respectively, with higher levels of ΣPCBs and ΣDDTs in the neonates and eggs than in muscle tissues. The ΣDDT residues in the studied Chinese alligators were at the high end of reported ranges from crocodilia around the world, and some results exceeded levels known to cause a female-biased sex ratio in crocodilians.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40124
4,4′-DDT solution, certified reference material, 5000 μg/mL in methanol
C14H9Cl5