EMAIL THIS PAGE TO A FRIEND
Acta chemica Scandinavica (Copenhagen, Denmark : 1989)
Facile synthesis of monoacetylated spermidines, illustrating selective deacetylation and application of a common precursor.
PMID 2488261
Abstract
The synthesis of all three monoacetylated spermidines is reported. N4-Acetylspermidine was obtained in four steps from spermidine via the triacetylated intermediate by selective deacetylation after exhaustive t-butoxycarbonylation as well as directly from a previously described protected precursor. N1-Acetylspermidine and N8-acetylspermidine were both obtained in four simple protection/deprotection steps from a common, selectively protected compound, thus illustrating the versatility of the latter.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3658
N8-Acetylspermidine dihydrochloride, analytical standard
C9H21N3O · 2HCl