EMAIL THIS PAGE TO A FRIEND
BMC Genomics
Nucleosome positioning and histone modifications define relationships between regulatory elements and nearby gene expression in breast epithelial cells.
PMID 24885402