EMAIL THIS PAGE TO A FRIEND
Archives of Pathology & Laboratory Medicine
ALK rearrangement in a large series of consecutive non-small cell lung cancers: comparison between a new immunohistochemical approach and fluorescence in situ hybridization for the screening of patients eligible for crizotinib treatment.
PMID 24885803