EMAIL THIS PAGE TO A FRIEND
Diagnostic pathology
The development of endometrial hyperplasia in aged PD-1-deficient female mice.
PMID 24886298
Abstract
Programmed death-1 (PD-1, Pdcd1)-deficient mice develop different types of autoimmune diseases depending on the mouse strain but its role in uterus development has not been reported. In this study, the expression of PD-1 and its ligands, PD-L1 and PD-L2, in uterine tissues from aged WT mice in a 129svEv-Brd background was analyzed by immunohistochemistry and the uterine morphology between WT and PD-1-/- mice was compared by hematoxylin and eosin staining. The aged PD-1-/- female mice in a 129svEv-Brd rather than Balb/c background develop endometrial hyperplasia. H&E staining showed an increase in the number of glands, neovascularization and an extremely large luminal cavity in aged PD-1-/- uteri. Immunohistochemical assay showed that the expression of PD-1 was observed in glandular/luminal epithelium and cells infiltrating the stroma. Fluorescent double staining demonstrated that PD-1 expresses on CD68+ macrophages, CD3+ T cells, CD16+ monocytes, CD56+ NK cells and CK-18+ epithelial cells, respectively. Additionally, PD-1 co-expresses with vascular endothelial growth factor (VEGF), and PD-1 deficiency resulted in an accumulation of glandular/luminal epithelium derived VEGF, which accelerates the expression of the proliferation-associated protein, proliferating cell nuclear antigen (PCNA), and thus potentially lead to epithelial proliferation in aged PD-1-/- uteri. These findings showed that PD-1 deficiency augments luminal epithelial cell proliferation probably through induced VEGF secretion, suggesting PD-1 plays an important role in controlling the growth and differentiation of the uterine epithelium. The virtual slide(s) for this article can be found here: http://www.diagnosticpathology.diagnomx.eu/vs/5809067461223905.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS4065
Anti-PD-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4067
Anti-PD-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900373
Anti-PDCD1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500763
Anti-PDCD1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400196
Anti-PDCD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA035981
Anti-PDCD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA013411
Anti-PDCD1LG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411458
Anti-PDCD1LG2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
V6627 Anti-Vascular Endothelial Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
V1253 Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3500122
Monoclonal Anti-PD-1 antibody produced in mouse, clone 7A11B1, purified immunoglobulin, buffered aqueous solution
SAB3500123
Monoclonal Anti-PD-1 antibody produced in mouse, clone 12A7D7, purified immunoglobulin, buffered aqueous solution
V4758 Monoclonal Anti-Vascular Endothelial Growth Factor antibody produced in mouse, clone 26503, purified immunoglobulin, lyophilized powder
SAB1402390
Monoclonal Anti-VEGF antibody produced in mouse, clone 3F7, purified immunoglobulin, buffered aqueous solution