EMAIL THIS PAGE TO A FRIEND
Acta Neuropathologica Communications
Ischemia induces different levels of hypoxia inducible factor-1? protein expression in interneurons and pyramidal neurons.
PMID 24887017