EMAIL THIS PAGE TO A FRIEND
Journal of Cardiovascular Pharmacology
Assessment of efficacy of proarrhythmia biomarkers in isolated rabbit hearts with attenuated repolarization reserve.
PMID 24887684
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000882
Adrenaline, European Pharmacopoeia (EP) Reference Standard
C9H13NO3