EMAIL THIS PAGE TO A FRIEND
European Journal of Haematology
Senescence of bone marrow mesenchymal stromal cells is accompanied by activation of p53/p21 pathway in myelodysplastic syndromes.
PMID 24889123