EMAIL THIS PAGE TO A FRIEND
Alcoholism, Clinical and Experimental Research
Contrasting influences of Drosophila white/mini-white on ethanol sensitivity in two different behavioral assays.
PMID 24890118