EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Metal-organic framework UiO-66 coated stainless steel fiber for solid-phase microextraction of phenols in water samples.
PMID 24891159
Abstract
Effective solid-phase microextraction (SPME) of polar phenols from water samples is usually difficult due to the strong interaction between polar phenols and aqueous matrix. Here, we report the fabrication of a metal-organic framework UiO-66 coated stainless steel fiber via physical adhesion for the SPME of polar phenols (phenol, o-cresol, p-cresol, 2,6-dimethylphenol, 2,4-dichlorophenol and 2,6-dichlorophenol) in water samples before gas chromatographic separation with flame ionic detection. Headspace SPME of 10mL sample solution with the fabricated UiO-66 coated fiber gave the enhancement factors of 160 (phenol) - 3769 (2,4-dichlorophenol), and the linear ranges of 1-1000μgL(-1) (2,6-dimethylphenol, 2,4-dichlorophenol and 2,6-dichlorophenol), 1-500μgL(-1) (o-cresol and p-cresol) and 5-500μgL(-1) (phenol). The detection limits ranged from 0.11μgL(-1) (2,6-dimethylphenol) to 1.23μgL(-1) (phenol). The precision (relative standard deviations, RSDs) for six replicate determinations of the analytes at 100μgL(-1) using a single UiO-66 coated fiber ranged from 2.8% to 6.2%. The fiber-to-fiber reproducibility (RSDs) for three parallel UiO-66 coated fibers varied from 5.9% to 10%. The recoveries obtained by spiking 5μgL(-1) of the phenols in the water samples ranged from 80% to 115%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
419141
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zr
403296
Zirconium, powder, −100 mesh
Zr
267724
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zr
267651
Zirconium, sponge, ≥99% trace metals basis
Zr
756385
Zirconium, powder, in water, -325 mesh, 99.5% trace metals basis (excluding Hf)
Zr
GF69451487
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zr
GF27504402
Zirconium, foil, 300x300mm, thickness 0.5mm, annealed, 99.2%
Zr
GF74069837
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zr
GF13629483
Zirconium, foil, 50x50mm, thickness 0.1mm, annealed, 99.2%
Zr
GF87053474
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zr
GF83603909
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zr
GF04051886
Zirconium, foil, 50x50mm, thickness 2.0mm, annealed, 99.2%
Zr
GF14815593
Zirconium, foil, 10mm disks, thickness 0.0125mm, 99.2%
Zr
GF58113019
Zirconium, foil, 10mm disks, thickness 0.015mm, 99.2%
Zr
GF67870468
Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%
Zr
GF58442070
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF54823642
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF75562869
Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%
Zr
GF55287221
Zirconium, foil, 10mm disks, thickness 0.02mm, 99.2%
Zr
GF38009044
Zirconium, foil, 10mm disks, thickness 0.05mm, annealed, 99.2%
Zr
GF89139594
Zirconium, foil, 10mm disks, thickness 0.075mm, annealed, 99.2%
Zr
GF22007595
Zirconium, foil, 10mm disks, thickness 0.125mm, annealed, 99.2%
Zr
GF50038039
Zirconium, foil, 10mm disks, thickness 0.25mm, annealed, 99.2%
Zr
GF03886443
Zirconium, foil, 10mm disks, thickness 0.5mm, annealed, 99.2%
Zr
GF09475027
Zirconium, foil, 15mm disks, thickness 0.009mm, 99.2%
Zr
GF51948872
Zirconium, foil, 15mm disks, thickness 0.0125mm, 99.2%
Zr
GF83808221
Zirconium, foil, 15mm disks, thickness 0.016mm, 99.2%
Zr
GF05259135
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zr
GF72812370
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zr
GF45357688
Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%
Zr