EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Capillary atmospheric pressure chemical ionization using liquid point electrodes.
PMID 24895256
Abstract
Atmospheric pressure chemical ionization (APCI) sources operated with point to plane DC discharges ('Coronas') frequently suffer from point electrode degradation and potentially lead to oxidation and/or fragmentation of the generated analyte ions. It is postulated that these adverse effects are caused by the interaction of these ions with the discharge chemistry as well as en route to the mass analyzer region. The corona discharge metal point electrode is replaced by the conically shaped liquid effluent evolving from a fused-silica capillary, which is analogous but not identical to the Taylor cone formation in electrospray ionization. The liquid consisting of either pure water or water containing 0.1 %V formic acid is fed via a nano-flow delivery stage at typical flow rates between 1-800 μL/h. The liquid flow is continuously replenishing the surface of the point electrode. The source is directly coupled to the inlet capillary of appropriate mass spectrometers, e.g., the Bruker Daltonics and Agilent varieties. The actively pumped liquid flow is supplying a constant amount of the reagent gas (H2O) to the corona region in the 20 ppmV to 30 %V range, leading to controlled, very stable operation of the source. The typical light emission observed for corona discharges is in very close proximity to the aqueous surface. Analyte protonation is the dominating ionization pathway. The degree of primary analyte fragmentation is extremely low. We have developed a novel atmospheric pressure chemical ionization source designed for the hyphenation of nano-flow liquid chromatography and gas chromatography with atmospheric pressure ionization mass spectrometry. The proposed reaction mechanism including the electrochemistry occurring in the source along with formation of protonated analyte molecules via collision-induced dissociation (CID) is in full accord with the experimental results. The system exhibits an extremely stable performance over prolonged operation times, sole generation of protonated molecules, and low degree of analyte ion fragmentation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93171
Diethylene glycol, BioUltra, ≥99.0% (GC)
C4H10O3
03128
Diethylene glycol, analytical standard
C4H10O3
32160
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
C4H10O3
PHR1045
Diethylene glycol
C4H10O3
1193265
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
C4H10O3
H26456
Diethylene glycol, ReagentPlus®, 99%
C4H10O3
295299
Dimethyl ether, ≥99%
C2H6O
61689
Methyl dodecanoate, analytical standard
C13H26O2
39937
Methyl laurate, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C13H26O2
234591
Methyl laurate, 99.5%
C13H26O2
07041
Methyl laurate, certified reference material, TraceCERT®
C13H26O2
W271500
Methyl laurate, ≥98%, FG
C13H26O2
W271511
Methyl laurate, natural, 98%, FG
C13H26O2
M3378
Methyl myristate, ≥99% (GC)
C15H30O2
55791
Methyl myristate, certified reference material, TraceCERT®
C15H30O2
70129
Methyl myristate, analytical standard
C15H30O2
W272205
Methyl myristate, ≥98%, FG
C15H30O2
P5177
Methyl palmitate, ≥99% (capillary GC)
C17H34O2
76159
Methyl palmitate, analytical standard
C17H34O2
W509531
Methyl palmitate, ≥97%
C17H34O2
S5376
Methyl stearate, ~99% (GC)
C19H38O2
75533
Methyl stearate, certified reference material, TraceCERT®
C19H38O2
85769
Methyl stearate, analytical standard
C19H38O2
W504807
Methyl stearate, ≥96%, FG
C19H38O2
M1770200
Methyl stearate, European Pharmacopoeia (EP) Reference Standard
C19H38O2