EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
Behavior of sartans (antihypertensive drugs) in wastewater treatment plants, their occurrence and risk for the aquatic environment.
PMID 24898294