EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Anti-inflammatory effects of Perilla frutescens leaf extract on lipopolysaccharide-stimulated RAW264.7 cells.
PMID 24898576
Abstract
Perilla leaves are widely used in Chinese herbal medicine and in Japanese herbal agents used to treat respiratory diseases. This study aimed to investigate the anti‑inflammatory effects and the underlying mechanisms of Perilla frutescens leaf extract (PLE). Murine macrophage RAW264.7 cells were used as a model. Cell viability and morphological changes were studied by the MTT assay and microscopy. mRNA expression of pro‑inflammatory mediators was assessed by both semi‑quantitative reverse transcription‑polymerase chain reactionxa0(RT‑PCR) and quantitative (q) RT‑PCR. Nitric oxidexa0(NO) and prostaglandinxa0E2 (PGE2) production were analyzed by the Griess test and sandwich enzyme‑linked immunosorbent assay (ELISA), respectively. The activation of kinase cascades was studied by immunoblotting. Our findings showed that PLE slightly affects cell viability, but alleviates LPS‑induced activation of RAW264.7 cells. Furthermore, PLE significantly reduced the LPS‑induced mRNA expression of the interleukin (IL)‑6, IL‑8, tumor necrosis factor‑αxa0(TNF‑α), cyclooxygenase‑2xa0(COX‑2) and inducible nitric oxide synthasexa0(iNOS), genes in a dose‑dependent manner. In addition, PLE reduced NO production and PGE2 secretion induced by LPS. PLE also inhibited activation of mitogen‑activated protein kinasesxa0(MAPKs), increased the cytosolic IκBα level, and reduced the level of nuclear factor (NF)‑κB. Taken together, these findings indicate that PLE significantly decreases the mRNA expression and protein production of pro‑inflammatory mediators, via the inhibition of extracellular‑signal‑regulated kinase (ERK)1/2, c‑Jun N‑terminal kinase (JNK), p38, as well as NF‑κB signaling in RAW264.7 cells stimulated with LPS.