EMAIL THIS PAGE TO A FRIEND
Journal of virology
Crucial roles of interleukin-7 in the development of T follicular helper cells and in the induction of humoral immunity.
PMID 24899182
Abstract
T follicular helper (Tfh) cells are specialized providers of cognate B cell help, which is important in promoting the induction of high-affinity antibody production in germinal centers (GCs). Interleukin-6 (IL-6) and IL-21 have been known to play important roles in Tfh cell differentiation. Here, we demonstrate that IL-7 plays a pivotal role in Tfh generation and GC formation in vivo, as treatment with anti-IL-7 neutralizing antibody markedly impaired the development of Tfh cells and IgG responses. Moreover, codelivery of mouse Fc-fused IL-7 (IL-7-mFc) with a vaccine enhanced the generation of GC B cells as well as Tfh cells but not other lineages of T helper cells, including Th1, Th2, and Th17 cells. Interestingly, a 6-fold-lower dose of an influenza virus vaccine codelivered with Fc-fused IL-7 induced higher antigen-specific and cross-reactive IgG titers than the vaccine alone in both mice and monkeys and led to markedly enhanced protection against heterologous influenza virus challenge in mice. Enhanced generation of Tfh cells by IL-7-mFc treatment was not significantly affected by the neutralization of IL-6 and IL-21, indicating an independent role of IL-7 on Tfh differentiation. Thus, IL-7 holds promise as a critical cytokine for selectively inducing Tfh cell generation and enhancing protective IgG responses. Here, we demonstrate for the first time that codelivery of Fc-fused IL-7 significantly increased influenza virus vaccine-induced antibody responses, accompanied by robust expansion of Tfh cells and GC B cells as well as enhanced GC formation. Furthermore, IL-7-mFc induced earlier and cross-reactive IgG responses, leading to striking protection against heterologous influenza virus challenge. These results suggest that Fc-fused IL-7 could be used for inducing strong and cross-protective humoral immunity against highly mutable viruses, such as HIV and hepatitis C virus, as well as influenza viruses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
237086
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
AlKO8S2 · 12H2O
31242
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
AlKO8S2 · 12H2O
RAB0316
Human IL-7 ELISA Kit, for serum, plasma, cell culture supernatant and urine
SRP3244 IL-7 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
SRP3266 IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
I4892 Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I5896 Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SAB4700111 Monoclonal Anti-CD19-Biotin antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB4700477 Monoclonal Anti-CD45-Biotin antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution
RAB0317
Mouse IL-7 ELISA Kit, for serum, plasma and cell culture supernatant
X1251
Xylazine hydrochloride, ≥99.0% (HPLC)
C12H16N2S · HCl
46995
Xylazine hydrochloride, VETRANAL®, analytical standard
C12H16N2S · HCl
X0075000
Xylazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C12H16N2S · HCl