EMAIL THIS PAGE TO A FRIEND
ACS medicinal chemistry letters
Metabolically Stable tert-Butyl Replacement.
PMID 24900702
Abstract
Susceptibility to metabolism is a common issue with the tert-butyl group on compounds of medicinal interest. We demonstrate an approach of removing all the fully sp(3) C-Hs from a tert-butyl group: replacing some C-Hs with C-Fs and increasing the s-character of the remaining C-Hs. This approach gave a trifluoromethylcyclopropyl group, which increased metabolic stability. Trifluoromethylcyclopropyl-containing analogues had consistently higher metabolic stability in vitro and in vivo compared to their tert-butyl-containing counterparts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
790184
Sodium 1-(trifluoromethyl)cyclopropanesulfinate
C4H4F3NaO2S