PloS one
The NOD-like receptor signalling pathway in Helicobacter pylori infection and related gastric cancer: a case-control study and gene expression analyses.
PMID 24901306
Abstract
Currently, it is well established that cancer arises in chronically inflamed tissue. A number of NOD-like receptors (NLRs) form inflammasomes, intracellular multiprotein complexes critical for generating mature pro-inflammatory cytokines (IL-1β and IL-18). As chronic inflammation of the gastric mucosa is a consequence of Helicobacter pylori infection, we investigated the role of genetic polymorphisms and expression of genes involved in the NLR signalling pathway in H. pylori infection and related gastric cancer (GC). Fifty-one genetic polymorphisms were genotyped in 310 ethnic Chinese (87 non-cardia GC cases and 223 controls with functional dyspepsia). In addition, gene expression of 84 molecules involved in the NLR signalling pathway was assessed in THP-1 cells challenged with two H. pylori strains, GC026 (GC) and 26695 (gastritis). CARD8-rs11672725, NLRP3-rs10754558, NLRP3-rs4612666, NLRP12-rs199475867 and NLRX1-rs10790286 showed significant associations with GC. On multivariate analysis, CARD8-rs11672725 remained a risk factor (OR: 4.80, 95% CI: 1.39-16.58). Further, NLRP12-rs2866112 increased the risk of H. pylori infection (OR: 2.13, 95% CI: 1.22-3.71). Statistical analyses assessing the joint effect of H. pylori infection and the selected polymorphisms revealed strong associations with GC (CARD8, NLRP3, CASP1 and NLRP12 polymorphisms). In gene expression analyses, five genes encoding NLRs were significantly regulated in H. pylori-challenged cells (NLRC4, NLRC5, NLRP9, NLRP12 and NLRX1). Interestingly, persistent up-regulation of NFKB1 with simultaneous down-regulation of NLRP12 and NLRX1 was observed in H. pylori GC026-challenged cells. Further, NF-κB target genes encoding pro-inflammatory cytokines, chemokines and molecules involved in carcinogenesis were markedly up-regulated in H. pylori GC026-challenged cells. Novel associations between polymorphisms in the NLR signalling pathway (CARD8, NLRP3, NLRP12, NLRX1, and CASP1) and GC were identified in Chinese individuals. Our genetic polymorphisms and gene expression results highlight the relevance of the NLR signalling pathway in gastric carcinogenesis and its close interaction with NF-κB.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3773
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
R2655
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
G2904
Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062
Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148
Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891
Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
I6135
Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016
Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513
Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888
Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
R1008
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
R3155
Monoclonal Anti-Rabbit IgG, Native−Peroxidase antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
R0761
Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid
85886
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
C21H39N7O12 · 1.5H2SO4
S1400000
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
C21H39N7O12 · 1.5H2O4S