PLoS ONE
Sinomenine sensitizes multidrug-resistant colon cancer cells (Caco-2) to doxorubicin by downregulation of MDR-1 expression.
PMID 24901713
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001445
Celecoxib, European Pharmacopoeia (EP) Reference Standard
C17H14F3N3O2S