EMAIL THIS PAGE TO A FRIEND
Alimentary pharmacology & therapeutics
Letter: Limitations of studies to evaluate the significance of anti-tumour necrosis factor serum levels in Crohn's disease.
PMID 24903436