EMAIL THIS PAGE TO A FRIEND
International journal of nanomedicine
Cinnamaldehyde and cinnamaldehyde-containing micelles induce relaxation of isolated porcine coronary arteries: role of nitric oxide and calcium.
PMID 24904214