EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Determination of isoniazid in pharmaceutical preparations by reaction with radiochloramine-B.
PMID 2490531
Abstract
A method has been developed for the determination of isoniazid (INH) at microgram level in pharmaceutical preparations based on the oxidation of isoniazid by radiochloramine-B. Interference of vitamin C has been overcome by precipitation as lead ascorbate and that of rifampicin and p-aminosalicylic acid by selective extraction. Amounts as low as 25 micrograms of INH can be determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
23265
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis
C6H5ClNNaO2S · xH2O