EMAIL THIS PAGE TO A FRIEND
Colloids and Surfaces. B, Biointerfaces
In vitro degradability, bioactivity and cell responses to mesoporous magnesium silicate for the induction of bone regeneration.
PMID 24905677