EMAIL THIS PAGE TO A FRIEND
Omics : A Journal of Integrative Biology
Lipidomics for clinical diagnosis: Dye-Assisted Laser Desorption/Ionization (DALDI) method for lipids detection in MALDI mass spectrometry imaging.
PMID 24905741