EMAIL THIS PAGE TO A FRIEND
Journal of cell science
The ubiquitin-conjugating enzymes UBE2N, UBE2L3 and UBE2D2/3 are essential for Parkin-dependent mitophagy.
PMID 24906799
Abstract
Depolarized mitochondria are degraded by mitophagy in a process that depends on the Parkinson's disease gene products PINK1 and Parkin. This is accompanied by ubiquitylation of several mitochondrial substrates. The roles of E2 ubiquitin-conjugating enzymes (UBE2) in mitophagy are poorly understood. Here, we investigate a set of UBE2 enzymes that might regulate Parkin-mediated mitophagy. Knockdown of the E2 enzymes UBE2N, UBE2L3 or UBE2D2 and UBE2D3 (UBE2D2/3) significantly reduced autophagic clearance of depolarized mitochondria. However, this did not interfere with mitochondrial PINK1 stabilization and Parkin translocation. UBE2N knockdown prevented specifically K63-linked ubiquitylation at mitochondrial sites. Nevertheless, polyubiquitin and p62 (officially known as SQSTM1) were still found on mitochondria after individual UBE2 knockdown. Knockdown of all of these UBE2s together significantly reduced mitochondrial polyubiquitylation and p62 recruitment. Moreover, reduced ubiquitylation of mitofusins, the mitochondrial import receptor subunits TOM20 and TOM70, the voltage-dependent anion channel protein 1 and Parkin was observed in cells silenced for all of these UBE2s. A version of Parkin with a mutation in the active site (C431S) failed to ubiquitylate these mitochondrial substrates even in the presence of UBE2s. We conclude that UBE2N, UBE2L3 and UBE2D2/3 synergistically contribute to Parkin-mediated mitophagy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306284
ANTI-E2EPF(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
P0075
Anti-PINK1 (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA001931
Anti-PINK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
P0076
Anti-PINK1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1411280
Anti-PINK1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3500459
Anti-TOM70 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500987
Anti-TOM70 antibody produced in rabbit, affinity isolated antibody
HPA011562
Anti-TOMM20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014589
Anti-TOMM70 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102622
Anti-UBE2D3 antibody produced in rabbit, affinity isolated antibody
AV43026
Anti-UBE2L3 (AB1) antibody produced in rabbit, affinity isolated antibody
AV43025
Anti-UBE2L3 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2103279
Anti-UBE2L3 antibody produced in rabbit, affinity isolated antibody
HPA045609
Anti-UBE2L3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102626
Anti-UBE2S antibody produced in rabbit, affinity isolated antibody
AV35122
Anti-VDAC1 antibody produced in rabbit, IgG fraction of antiserum
SAB3300002
Monoclonal Anti-c-Myc antibody produced in mouse, clone 1A6, purified immunoglobulin, buffered aqueous glycerol solution
WH0002965M1
Monoclonal Anti-GTF2H1 antibody produced in mouse, clone 1F12-1B5, purified immunoglobulin, buffered aqueous solution
WH0055669M4
Monoclonal Anti-MFN1 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
WH0009927M3
Monoclonal Anti-MFN2 antibody produced in mouse, clone 4H8, purified immunoglobulin, buffered aqueous solution
M9073
Monoclonal Anti-Mitofusin 2 antibody produced in mouse, ~1.0 mg/mL, clone Mito-2, purified immunoglobulin, buffered aqueous solution
WH0004609M2
Monoclonal Anti-MYC antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
WH0023636M2
Monoclonal Anti-NUP62 antibody produced in mouse, clone 2D3, purified immunoglobulin, buffered aqueous solution