EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Profile of secreted hydrolases, associated proteins, and SlpA in Thermoanaerobacterium saccharolyticum during the degradation of hemicellulose.
PMID 24907337