EMAIL THIS PAGE TO A FRIEND
Steroids
Epigenetic modifier-induced biosynthesis of novel acetylenic sterols from Cladosporium colocasiae.
PMID 24907456
Abstract
The addition of an HDAC inhibitor, suberoylanilide hydroxamic acid (SBHA), to the culture medium of Cladosporium colocasiae, dramatically altered its metabolic profiles. Analysis of the culture broth extract led to the isolation of two new acetylenic sterols (1-2). The isolated compounds were further evaluated for their cytotoxic and antibacterial activities. Compound 1 showed activity against Bacillus subtilis, affording a zone of inhibition of 12mm at 100μg/disk. However, none of them showed noticeable growth inhibitory effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
390585
Suberohydroxamic acid, 95%
C8H16N2O4