EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Vigabatrin in dried plasma spots: validation of a novel LC-MS/MS method and application to clinical practice.
PMID 24908379