EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Chronic impact of sulfamethoxazole on acetate utilization kinetics and population dynamics of fast growing microbial culture.
PMID 24908607
Abstract
The study evaluated the chronic impact of sulfamethoxazole on metabolic activities of fast growing microbial culture. It focused on changes induced on utilization kinetics of acetate and composition of the microbial community. The experiments involved a fill and draw reactor, fed with acetate and continuous sulfamethoxazole dosing of 50 mg/L. The evaluation relied on model evaluation of the oxygen uptake rate profiles, with parallel assessment of microbial community structure by 454-pyrosequencing. Continuous sulfamethoxazole dosing inflicted a retardation effect on acetate utilization in a way commonly interpreted as competitive inhibition, blocked substrate storage and accelerated endogenous respiration. A fraction of acetate was utilized at a much lower rate with partial biodegradation of sulfamethoxazole. Results of pyrosequencing with a replacement mechanism within a richer more diversified microbial culture, through inactivation of vulnerable fractions in favor of species resistant to antibiotic, which made them capable of surviving and competing even with a slower metabolic response.