EMAIL THIS PAGE TO A FRIEND
Journal of the European Academy of Dermatology and Venereology : JEADV
Efficacy and safety of topical glycopyrrolate in patients with facial hyperhidrosis: a randomized, multicentre, double-blinded, placebo-controlled, split-face study.
PMID 24909188
Abstract
Although facial hyperhidrosis has been frequently associated with a diminished quality of life, various conservative modalities for its management are still far from satisfactory. To evaluate the antiperspirant efficacy and safety of the topical glycopyrrolate on facial hyperhidrosis at specified posttreatment intervals. Thirty-nine patients with facial hyperhidrosis were enrolled and treated with 2% topical glycopyrrolate on one-half of the forehead, whereas the other half of the forehead was treated with a placebo. All patients applied topical glycopyrrolate or placebo once a day for nine successive days. Each evaluation included weighing sweat and assessing the Hyperhidrosis Disease Severity Scale (HDSS) score and any adverse effects. Compared with the placebo-treated sides, topical glycopyrrolate-treated sides showed a reduction in the rate of sweat production at the forehead of 25.16 ± 10.30% (mean ± SD) at 90 min after the first application (day 1), 29.63 ± 7.74% at 24 h after the first application (day 2) and 36.68 ± 11.41% at 24 h after eight additional successive daily applications (day 10) (all P < 0.025). There was a little more decrease in HDSS score with the topical glycopyrrolate-treated half of the forehead, but the difference was not statistically significant (P > 0.025). No serious adverse events were reported during the course of this study. Only one patient developed a transient headache after treatment. Topical glycopyrrolate application appears to be significantly effective and safe in reducing excessive facial perspiration.