EMAIL THIS PAGE TO A FRIEND
Clinical and experimental pharmacology & physiology
Inhibitory action of naphtho[1,2-b]furan-4,5-dione on hepatocyte growth factor-induced migration and invasion of MDA-MB-231 cells: mechanisms of action.
PMID 24909202
Abstract
Naphtho[1,2-b]furan-4,5-dione (NFD), a bioactive component of Avicennia marina, has been shown to exhibit anticancer activity. The aim of the present study was to explore the effect of NFD on hepatocyte growth factor (HGF)-induced cell migration and invasion of MDA-MB-231 human breast cancer cells, as well as the underlying mechanism of action. Cell viability was determined using the 3-(4,5-dimethyl-2 thiazoyl)-2,5-diphenyl-2H-tetrazolium bromide assay, western blot analysis was used to measure protein expression and cell migration and invasion were evaluated by the cell wound healing assay, Boyden chamber assay and gelatin zymography. When cells were treated with non-toxic concentrations of NFD (1-3 μmol/L, 24 h), NFD concentration-dependently inhibited HGF-promoted cell migration and invasion. Simultaneously, NFD efficiently suppressed c-Met phosphorylation and downstream activation of phosphatidylinositol 3-kinase (PI3K) and Akt. In addition, NFD inhibited the phosphorylation of IκB kinases and IκBα and nuclear translocation of nuclear factor (NF)-κB, as well as matrix metalloproteinase (MMP)-9 activity. Furthermore, the c-Met inhibitor PHA665752 (10 μmol/L) inhibited HGF-induced MMP-9 expression, cell migration and invasion, as well as the activation of PI3K/Akt, suggesting that PI3K/Akt activation occur downstream of c-Met activation. In conclusion, the results of the present study suggest that NFD inhibits HGF-induced invasion and migration of MDA-MB-231 cells via HGF- and/or c-Met-mediated PI3K/Akt and NF-κB signalling pathways, leading to downregulation of MMP-9 expression and cell migration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40584
Anti-NCL (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1409341
Anti-NCL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA023981
Anti-NCL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410823
Anti-NCL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501861
Anti-NFASC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008832
Anti-NFASC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
N2662
Anti-Nucleolin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504017
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
SAB4300043
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504749
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4503852
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
SAB4300260
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
P3862 Anti-phospho-PKB (pThr308) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
GW21071 Anti-PI3K antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2701957
Anti-PIK3CA antibody produced in rabbit
HPA001216
Anti-PIK3R1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
H1404 Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
H5791 Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
SRP6014 HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
SRP6166
HGF human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
WH0005290M1
Monoclonal Anti-PIK3CA antibody produced in mouse, clone 3G3, purified immunoglobulin, buffered aqueous solution
SAB5300225
Monoclonal Anti-PIK3CA antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
SAB1404206
Monoclonal Anti-PIK3R1 antibody produced in mouse, clone 1C10, purified immunoglobulin, buffered aqueous solution
WH0005295M1
Monoclonal Anti-PIK3R1 antibody produced in mouse, clone 3A10, purified immunoglobulin, buffered aqueous solution
AV40583
Anti-NCL (AB1) antibody produced in rabbit, affinity isolated antibody
SAB4504015
Anti-phospho-Akt (pSer246) antibody produced in rabbit, affinity isolated antibody