EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Analysis of endocrine disrupters and related compounds in sediments and sewage sludge using on-line turbulent flow chromatography-liquid chromatography-tandem mass spectrometry.
PMID 24909438
Abstract
A novel fully automated method based on dual column switching using turbulent flow chromatography followed by liquid chromatography coupled to tandem mass spectrometry (TFC-LC-MS/MS) was applied for the determination of endocrine disruptors (EDCs) and related compounds in sediment and sewage sludge samples. This method allows the unequivocal identification and quantification of the most relevant environmental EDCs such as natural and synthetic estrogens and their conjugates, antimicrobials, parabens, bisphenol A (BPA), alkylphenolic compounds, benzotriazoles, and organophosphorus flame retardants, minimizing time of analysis and alleviating matrix effects. Applying this technique, after the extraction of the target compounds by pressurized liquid extraction (PLE), sediment and sewage sludge extracts were directly injected to the chromatographic system and the analytes were concentrated into the clean-up loading column. Using six-port switching system, the analytes were transferred to the analytical column for subsequent detection by MS-MS (QqQ). In order to optimize this multiplexing system, a comparative study employing six types of TurboFlow™ columns, with different chemical modifications, was performed to achieve the maximum retention of analytes and best elimination of matrix components. Using the optimized protocol low limits of quantification (LOQs) were obtained ranging from 0.0083 to 1.6 ng/g for sediment samples and from 0.10 to 125 ng/g for sewage sludge samples (except for alkylphenol monoethoxylate). The method was used to evaluate the presence and fate of target EDCs in the Ebro River which is the most important river in Spain with intensive agricultural and industrial activities in the basin that contribute to deteriorating soil and water quality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E4876
17α-Ethynylestradiol, ≥98%
C20H24O2
46263
17α-Ethynylestradiol, VETRANAL®, analytical standard
C20H24O2
290823
4-tert-Octylphenol, 97%
C14H22O
442858
4-tert-Octylphenol, analytical standard
C14H22O
32467
4-tert-Octylphenol solution, 1000 μg/mL in acetone, analytical standard
C14H22O
B11400
Benzotriazole, ReagentPlus®, 99%
C6H5N3
76457
Benzotriazole, analytical standard
C6H5N3
380709
Benzyl 4-hydroxybenzoate, 99%
C14H12O3
D4628
Diethylstilbestrol, ≥99% (HPLC)
C18H20O2
46207
Diethylstilbestrol, VETRANAL®, analytical standard
C18H20O2
D1500000
Diethylstilbestrol, European Pharmacopoeia (EP) Reference Standard
C18H20O2
218944
Diethylstilbestrol, mixture of cis and trans, 97%
C18H20O2
1250008
Estradiol, United States Pharmacopeia (USP) Reference Standard
C18H24O2
E1253
Estriol, ≥97%
C18H24O3
E1680000
Estriol, European Pharmacopoeia (EP) Reference Standard
C18H24O3
46565
Estriol, VETRANAL®, analytical standard
C18H24O3
E1149
Estriol, meets USP testing specifications
C18H24O3
E9750
Estrone, ≥99%
C18H22O2
46573
Estrone, VETRANAL®, analytical standard
C18H22O2
PHR1535
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
C18H22O2
1255001
Estrone, United States Pharmacopeia (USP) Reference Standard
C18H22O2
E1700000
Estrone, European Pharmacopoeia (EP) Reference Standard
C18H22O2
1260001
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
C20H24O2
PHR1480
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
C20H24O2
E1900000
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
C20H24O2
BP421
Ethinylestradiol, British Pharmacopoeia (BP) Assay Standard
C20H24O2
Y0001124
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
C20H24O2
111988
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
C9H10O3
E2425000
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C9H10O3
112291
Ethyl salicylate, 99%
C9H10O3