EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Histone deacetylase inhibition destabilizes the multi-potent state of uncommitted adipose-derived mesenchymal stromal cells.
PMID 24912092
Abstract
Human adipose-derived mesenchymal stromal cells (AMSCs) grown in platelet lysate are promising agents for therapeutic tissue regeneration. Here, we investigated whether manipulation of epigenetic events by the clinically relevant histone deacetylase inhibitor suberoylanilide hydroxamic acid (SAHA) alters differentiation of AMSCs. The multipotency of AMSCs was validated by their ability to differentiate into osteogenic, chondrogenic, and adipogenic lineages. High-throughput RNA sequencing and RT-qPCR established that human histone deacetylases (HDAC1 to HDAC11, and SIRT1 to SIRT7) are differentially expressed in AMSCs. SAHA induces hyper-acetylation of histone H3 and H4, stimulates protein expression of the HDAC-responsive gene SLC9A3R1/NHERF1 and modulates the AKT/FOXO1 pathway. Biologically, SAHA interferes with osteogenic, chondrogenic and adipogenic lineage commitment of multipotent AMSCs. Mechanistically, SAHA-induced loss of differentiation potential of uncommitted AMSCs correlates with multiple changes in the expression of principal transcription factors that control mesenchymal or pluripotent states. We propose that SAHA destabilizes the multi-potent epigenetic state of uncommitted human AMSCs by hyper-acetylation and perturbation of key transcription factor pathways. Furthermore, AMSCs grown in platelet lysate may provide a useful biological model for screening of new HDAC inhibitors that control the biological fate of human mesenchymal stromal cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50020
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
C3H7Na2O6P · 5H2O