EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Polyphenols from the stems of Morus alba and their inhibitory activity against nitric oxide production by lipopolysaccharide-activated microglia.
PMID 24912117
Abstract
Neuroinflammatory processes are involved in the pathogenesis of many neurodegenerative disorders. Microglial cells, the main immune cells of the central nervous system, represent a target of interest to search for naturally occurring anti-inflammatory products. In this study, we evaluated the anti-inflammatory properties of polyphenols obtained from the stems of Morus alba. This edible species, known as white mulberry, is frequently studied because of its traditional use in Asian medicine and its richness in different types of polyphenols, some of which are known to be phytoalexins. One new coumarin glycoside, isoscopoletin 6-(6-O-β-apiofuranosyl-β-glucopyranoside) (1) was mainly isolated by CPC (centrifugal partition chromatography) from this plant, together with seven known polyphenols (2-8). Their structures were established on the basis of spectroscopic analyses including extensive 2D NMR studies. The eight isolated compounds were evaluated for their inhibitory activities on nitric oxide (NO) production in lipopolysaccharide (LPS)-induced BV-2 microglial cells. The absence of cell toxicity is checked by a MTT assay.