EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Genome-wide RNAi screen identifies the Parkinson disease GWAS risk locus SREBF1 as a regulator of mitophagy.
PMID 24912190