EMAIL THIS PAGE TO A FRIEND
Molecular & cellular proteomics : MCP
Early targets of miR-34a in neuroblastoma.
PMID 24912852
Abstract
Several genes encoding for proteins involved in proliferation, invasion, and apoptosis are known to be direct miR-34a targets. Here, we used proteomics to screen for targets of miR-34a in neuroblastoma (NBL), a childhood cancer that originates from precursor cells of the sympathetic nervous system. We examined the effect of miR-34a overexpression using a tetracycline inducible system in two NBL cell lines (SHEP and SH-SY5Y) at early time points of expression (6, 12, and 24 h). Proteome analysis using post-metabolic labeling led to the identification of 2,082 proteins, and among these 186 were regulated (112 proteins down-regulated and 74 up-regulated). Prediction of miR-34a targets via bioinformatics showed that 32 transcripts held miR-34a seed sequences in their 3'-UTR. By combining the proteomics data with Kaplan Meier gene-expression studies, we identified seven new gene products (ALG13, TIMM13, TGM2, ABCF2, CTCF, Ki67, and LYAR) that were correlated with worse clinical outcomes. These were further validated in vitro by 3'-UTR seed sequence regulation. In addition, Michigan Molecular Interactions searches indicated that together these proteins affect signaling pathways that regulate cell cycle and proliferation, focal adhesions, and other cellular properties that overall enhance tumor progression (including signaling pathways such as TGF-β, WNT, MAPK, and FAK). In conclusion, proteome analysis has here identified early targets of miR-34a with relevance to NBL tumorigenesis. Along with the results of previous studies, our data strongly suggest miR-34a as a useful tool for improving the chance of therapeutic success with NBL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1304399
ANTI-ALG13 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1401832
Anti-ALG13 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002853
Anti-ALG13 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400621
Anti-LYAR antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1400622
Anti-LYAR antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB4502334
Anti-LYAR antibody produced in rabbit, affinity isolated antibody
HPA035881
Anti-LYAR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV47471
Anti-TGM2 (AB1) antibody produced in rabbit, affinity isolated antibody
AV47472
Anti-TGM2 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2501034
Anti-TGM2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502010
Anti-TGM2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406528
Anti-TGM2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029518
Anti-TGM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA021019
Anti-TGM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA048985
Anti-TIMM13 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107083
Anti-TIMM13 antibody produced in rabbit, affinity isolated antibody
SAB1403187
Monoclonal Anti-TIMM13 antibody produced in mouse, clone 4F4, ascites fluid
SAB2106035
Anti-LYAR antibody produced in rabbit, affinity isolated antibody
SAB1404467
Monoclonal Anti-TGM2, (N-terminal) antibody produced in mouse, clone 1A8, purified immunoglobulin, buffered aqueous solution
SAB2107240
Anti-LYAR antibody produced in rabbit, affinity isolated antibody