EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research. Part B, Applied Biomaterials
Encapsulation of porcine pancreatic islets within an immunoprotective capsule comprising methacrylated glycol chitosan and alginate.
PMID 24915784