EMAIL THIS PAGE TO A FRIEND
Clinical orthopaedics and related research
Reply to the letter to the editor: symptomatic pulmonary embolus after joint arthroplasty: stratification of risk factors.
PMID 24916711