EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Dual protonophore-chitinase inhibitors dramatically affect O. volvulus molting.
PMID 24918716
Abstract
The L3-stage-specific chitinase OvCHT1 has been implicated in the development of Onchocerca volvulus, the causative agent of onchocerciasis. Closantel, a known anthelmintic drug, was previously discovered as a potent and specific OvCHT1 inhibitor. As closantel is also a known protonophore, we performed a simple scaffold modulation to map out the structural features that are relevant for its individual or dual biochemical roles. Furthermore, we present that either OvCHT1 inhibition or protonophoric activity was capable of affecting O. volvulus L3 molting and that the presence of both activities in a single molecule yielded more potent inhibition of the nematode's developmental process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34093
Closantel, PESTANAL®, analytical standard
C22H14Cl2I2N2O2