EMAIL THIS PAGE TO A FRIEND
Organic & Biomolecular Chemistry
Outstanding effects on antithrombin activity of modified TBA diastereomers containing an optically pure acyclic nucleotide analogue.
PMID 24920241