EMAIL THIS PAGE TO A FRIEND
The American journal of surgical pathology
Risk of secondary malignancy (including breast) in patients with mismatch-repair protein deficiency.
PMID 24921635
Abstract
Lynch syndrome (LS) is an autosomal dominant inherited disease that is associated with an increased risk for colorectal and endometrial cancer due to germline mutations in mismatch-repair (MMR) genes. Whereas primary tumors in this syndrome are widely recognized, the relative risk(s) of secondary malignancies, particularly breast cancer, in LS patients are still poorly characterized. To provide an improved assessment of these risks, MMR status was evaluated in secondary tumors from a series of patients with index tumors of known MMR status (both proficient and deficient). A total of 1252 tumors (index tumors) and all secondary malignancies were tested for MMR by immunohistochemistry (MSH2, MSH6, MLH1, PMS2) between 1992 and 2013. Tumors with MLH1/PMS2 deficiency were tested for hypermethylation or BRAF mutation, when appropriate. Of the 1252 index tumors, 162 were MMR deficient (dMMR), and, of that subset, 32 secondary tumors were identified (19.7%). In contrast, 80 secondary tumors were identified in the proficient (intact) group (7.3%). Although secondary malignancies were more common in the dMMR group (P=0.0001), there was no trend in tumor type. Specifically, breast cancer was not overly represented in the dMMR group. When secondary tumors had dMMR, they were more likely to have deficiency in MSH2/MSH6 than in MLH1/PMS2 (P=0.01). Of the patients with tumors exhibiting dMMR, women were more likely to have a dMMR secondary tumor in this series (P=0.0001); however, breast cancer was not overly represented, and our study provides no evidence that it is more frequent in LS. MSH2/MSH6 deficiency is more commonly associated with a secondary tumor compared with MLH1/PMS2 deficiency, when methylation/BRAF status is taken into account.