EMAIL THIS PAGE TO A FRIEND
Arthritis Research & Therapy
Acid-sensing ion channel 3 decreases phosphorylation of extracellular signal-regulated kinases and induces synoviocyte cell death by increasing intracellular calcium.
PMID 24923411
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1530
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
C20H20N2O3