EMAIL THIS PAGE TO A FRIEND
Nature communications
RNA-binding proteins regulate the expression of the immune activating ligand MICB.
PMID 24924487
Abstract
The recognition of stress-induced ligands by the activating receptor NKG2D expressed on cytotoxic lymphocytes is crucial for the prevention and containment of various diseases and is also one of the best-studied examples of how danger is sensed by the immune system. Still, however, the mechanisms leading to the expression of the NKG2D ligands are far from being completely understood. Here, we use an unbiased and systematic RNA pull-down approach combined with mass spectrometry to identify six RNA-binding proteins (RBPs) that bind and regulate the expression of MICB, one of the major stress-induced ligands of NKG2D. We further demonstrate that at least two of the identified RBPs function during genotoxic stress. Our data provide insights into stress recognition and hopefully open new therapeutic venues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-225
Anti-c-Jun Antibody, Detect c-Jun, also known as transcription factor AP-1, with Anti-c-Jun rabbit polyclonal antiserum, Cat. No. 06-225, that has been demonstrated to work in Immunocytochemistry, Immunoprecipitation, and Western blotting.
05-594
Anti-HSP90 Antibody, This Anti-HSP90 Antibody is validated for use in IP, WB for the detection of HSP90.
J104 Anti-c-Jun antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4500141
Anti-CELF-1 antibody produced in rabbit, affinity isolated antibody
SAB2701623
Anti-celf1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1408786
Anti-CUGBP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401532
Anti-CUGBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103977
Anti-CUGBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1408970
Anti-ELAVL1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701888
Anti-elavl1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1410329
Anti-ELAVL1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500531
Anti-IGF2BP2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104501
Anti-IGF2BP2 antibody produced in rabbit, affinity isolated antibody
HPA035145
Anti-IGF2BP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300307
Anti-JUN (Ab-170) antibody produced in rabbit, affinity isolated antibody
SAB4300308
Anti-JUN (Ab-239) antibody produced in rabbit, affinity isolated antibody
SAB4300309
Anti-JUN (Ab-243) antibody produced in rabbit, affinity isolated antibody
SAB4300286
Anti-JUN (Ab-63) antibody produced in rabbit, affinity isolated antibody
SAB4300288
Anti-JUN (Ab-73) antibody produced in rabbit, affinity isolated antibody
SAB4300305
Anti-JUN (Ab-91) antibody produced in rabbit, affinity isolated antibody
SAB4300306
Anti-JUN (Ab-93) antibody produced in rabbit, affinity isolated antibody
AV100614
Anti-JUN (AB1) antibody produced in rabbit, affinity isolated antibody
AV100708
Anti-JUN (AB2) antibody produced in rabbit, affinity isolated antibody
AV100911
Anti-JUN (AB3) antibody produced in rabbit, affinity isolated antibody
SAB1406055
Anti-JUN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38133
Anti-JUN antibody produced in rabbit, IgG fraction of antiserum
SAB2103656
Anti-JUN antibody produced in rabbit, affinity isolated antibody
AV40922
Anti-MATR3 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV40923
Anti-MATR3 (AB2) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
HPA036564
Anti-MATR3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution