EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance imaging : JMRI
Clinical utility of endorectal MRI-guided prostate biopsy: preliminary experience.
PMID 24924999
Abstract
To investigate the potential clinical utility of endorectal MRI-guided biopsy in patients with known or suspected prostate cancer. We prospectively recruited 24 men with known or suspected prostate cancer in whom MRI-guided biopsy was clinically requested after multiparametric endorectal MRI showed one or more appropriate targets. One to six 18-gauge biopsy cores were obtained from each patient. Transrectal ultrasound guided biopsy results and post MRI-guided biopsy complications were also recorded. MRI-guided biopsy was positive in 5 of 7 patients with suspected prostate cancer (including 2 of 4 with prior negative ultrasound-guided biopsies), in 8 of 12 with known untreated prostate cancer (including 5 where MRI-guided biopsy demonstrated a higher Gleason score than ultrasound guided biopsy results), and in 3 of 5 with treated cancer. MRI-guided biopsies had a significantly higher maximum percentage of cancer in positive cores when compared with ultrasound guided biopsy (mean of 37 ± 8% versus 13 ± 4%; P = 0.01). No serious postbiopsy complications occurred. Our preliminary experience suggests endorectal MRI-guided biopsy may safely contribute to the management of patients with known or suspected prostate cancer by making a new diagnosis of malignancy, upgrading previously diagnosed disease, or diagnosing local recurrence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295736
Carbon tetrafluoride, 99.9%
CF4