EMAIL THIS PAGE TO A FRIEND
International journal of cosmetic science
Exogenous connective tissue growth factor preserves the hair-inductive ability of human dermal papilla cells.
PMID 24925376
Abstract
Connective tissue growth factor influences human dermal papilla cells' hair inductive ability through several signaling pathways.