EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Resolvin E1 reduces hepatic fibrosis in mice with
PMID 24926330
Abstract
The aim of this study was to investigate whether resolvin E1 (RvE1) protects against hepatic fibrosis in a murine model of liver fibrosis induced by