EMAIL THIS PAGE TO A FRIEND
Environment international
Australia's leading public health body delays action on the revision of the public health goal for blood lead exposures.
PMID 24927499
Abstract
Globally, childhood blood lead levels have fallen precipitously in developed countries since the 1970s following action by international bodies such as the WHO and Food and Agricultural Organization (FAO) of the United Nations. These reductions have been affected by the activities of national agencies such as the US EPA and US Centers for Disease Control and Prevention in the establishment of air lead and blood lead standards, the introduction of legislation to remove lead from petrol, paint and consumer products and tighter restrictions on lead emissions. The outcome of recent major international reviews of research into the effects of low-level lead exposures (e.g. by WHO, USA health and environmental agencies, German and Canadian health bodies) has resulted in recommendations to reduce and eliminate lead exposures. By contrast, Australian policy responses to the incontrovertible evidence that adverse neurocognitive and behavioural effects that occur at levels well below the current national goal of 10μg/dL have stalled. The delayed response by Australia occurs at a time when blood lead levels in two of Australia's three primary lead mining and smelting cities: Port Pirie, South Australia and Broken Hill, New South Wales, are rising. In the third city, Mount Isa, Queensland, there is still no systematic, annual testing of childhood blood lead values. This is despite the fact that Mount Isa has the highest lead (and other toxic metals such as cadmium and arsenic) emissions to the environment (120tonnes of lead in 2011/12) from any single point source in Australia. It is clear that both state and national policy approaches to the ongoing risks of lead exposure need to be revised urgently and in line with contemporary international standards. Recommended changes should include a new lower blood lead intervention level of no more than 5μg/dL, with a national goal for all children under 5years of age to have a blood lead level of below 1μg/dL by 2020. In order to achieve any new lower exposure goals other relevant lead standards including air, dust, soil and water must also be revised downwards.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF17721637
Cadmium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF76768914
Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF94010453
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF90633652
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cd
GF06836238
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cd
GF71408763
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF78263411
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF84554613
Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF19008817
Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF20958947
Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF15879954
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF72851182
Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF61336997
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cd