EMAIL THIS PAGE TO A FRIEND
Journal of Parkinson's Disease
Metabolite and peptide levels in plasma and CSF differentiating healthy controls from patients with newly diagnosed Parkinson's disease.
PMID 24927756
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45840
Acetic anhydride
C4H6O3
92360
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
C3H9ClSi
89595
Chlorotrimethylsilane, for GC derivatization, ≥99.0% (GC)
C3H9ClSi
G0355000
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
C5H9NO4
M3128
Myristic acid, Sigma Grade, ≥99%
C14H28O2
70079
Myristic acid, analytical standard
C14H28O2
70082
Myristic acid, ≥98.0% (GC)
C14H28O2
W276405
Myristic acid, ≥95%, FCC, FG
C14H28O2
W276413
Myristic acid, natural, ≥98.5%, FG
C14H28O2
130087
Myristic acid, 95%
C14H28O2
PHR1124
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C14H28O2
M0789
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥97.0% (GC)
C5H3F6NO2
M9513
N-Methyl-bis(trifluoroacetamide), ~95% (capillary GC)
C5H3F6NO2
65943
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥99.0% (GC)
C5H3F6NO2
43051
Palmitic acid, certified reference material, TraceCERT®
C16H32O2
1492007
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
C16H32O2
76119
Palmitic acid, analytical standard
C16H32O2
76122
Palmitic acid, purum, ≥98.0% (GC)
C16H32O2
PHR1120
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C16H32O2
P0090000
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
C16H32O2
P3350000
Proline, European Pharmacopoeia (EP) Reference Standard
C5H9NO2
S5922
Salicylic acid, BioXtra, ≥99.0%
C7H6O3
S7401
Salicylic acid, plant cell culture tested
C7H6O3
84210
Salicylic acid, puriss. p.a., ≥99.0% (T)
C7H6O3
27301
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
C7H6O3
1609002
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O3
84211
Salicylic acid, tested according to Ph.Eur.
C7H6O3
PHR1013
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O3
105910
Salicylic acid, ReagentPlus®, ≥99%
C7H6O3
W398500
Salicylic acid, ≥99%, FG
C7H6O3